Anti-REL Rabbit polyclonal antibody

ABCAAB19464-50EA 0 EUR
ABCAAB19464-50
Anti-REL Rabbit polyclonal antibody
Type: Primary
Antigen: REL (v-rel avian reticuloendotheliosis viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR