Anti-OPN4 Rabbit Polyclonal Antibody
Proveedor: Abcam
ABCAAB19383-100EA 550 EUR
ABCAAB19383-100
Anti-OPN4 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: OPN4 (opsin 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat