Anti-fetal hemoglobin Sheep Polyclonal Antibody (FITC (Fluorescein))

Proveedor: Abcam
ABCAAB19365-1EA 739 EUR
ABCAAB19365-1
Anti-fetal hemoglobin Sheep Polyclonal Antibody (FITC (Fluorescein))
Anticuerpos
Type: Primary
Antigen: fetal hemoglobin
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR