Anti-PLLP Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB19309-100EA 0 EUR
ABCAAB19309-100
Anti-PLLP Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PLLP (plasmolipin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR