Anti-PHAP Rabbit polyclonal antibody

ABCAAB19281-50EA 604 EUR
ABCAAB19281-50
Anti-PHAP Rabbit polyclonal antibody
Type: Primary
Antigen: PHAP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR