Anti-BLVRA Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB19260-50EA 739 EUR
ABCAAB19260-50
Anti-BLVRA Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: BLVRA (biliverdin reductase A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR