Anti-RFC4 Mouse monoclonal antibody [clone: 1320]

ABCAAB19259-50EA 588 EUR
ABCAAB19259-50
Anti-RFC4 Mouse monoclonal antibody [clone: 1320]
Type: Primary
Antigen: RFC4 (replication factor C (activator 1) 4, 37kDa)
Clonality: monoclonal
Clone: 1320
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR