Anti-HIV1 p24 Mouse monoclonal antibody [clone: 473]

ABCAAB19054-250EA 611 EUR
ABCAAB19054-250
Anti-HIV1 p24 Mouse monoclonal antibody [clone: 473]
Type: Primary
Antigen: HIV1 p24
Clonality: monoclonal
Clone: 473
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR