Anti-Pericentrin 1 Goat Polyclonal Antibody

Proveedor: Abcam
ABCAAB19044-100EA 799 EUR
ABCAAB19044-100
Anti-Pericentrin 1 Goat Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Pericentrin 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR