Anti-CETP Rabbit polyclonal antibody unconjugated
Proveedor: Abcam
ABCAAB19012-100EA 530 EUR
ABCAAB19012-100
Anti-CETP Rabbit polyclonal antibody unconjugated
Anticuerpos
Type: Primary
Antigen: CETP (cholesteryl ester transfer protein, plasma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now