Anti-LCK Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB18896-100EA 763 EUR
ABCAAB18896-100
Anti-LCK Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: LCK (lymphocyte-specific protein tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR