Anti-SYN1 Rabbit Polyclonal Antibody
Proveedor: Abcam
ABCAAB18814-100EA 570 EUR
ABCAAB18814-100
Anti-SYN1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: SYN1 (synapsin I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat