Anti-Hepatitis C Virus NS3 Mouse monoclonal antibody [clone: 5]

ABCAAB18666-250EA 0 EUR
ABCAAB18666-250
Anti-Hepatitis C Virus NS3 Mouse monoclonal antibody [clone: 5]
Type: Primary
Antigen: Hepatitis C Virus NS3
Clonality: monoclonal
Clone: 5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR