Anti-V5 Tag Rabbit Polyclonal Antibody (Biotin)
Proveedor: Abcam
ABCAAB18617-250EA 550 EUR
ABCAAB18617-250
Anti-V5 Tag Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Type: Primary
Antigen: V5 Tag (V5 tag)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now