Anti-Glu-Glu tag Rabbit polyclonal antibody

ABCAAB18585-200EA 604 EUR
ABCAAB18585-200
Anti-Glu-Glu tag Rabbit polyclonal antibody
Type: Primary
Antigen: Glu-Glu tag
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR