Anti-FSBA Mouse Monoclonal Antibody [clone: 2F5/5]

Proveedor: Abcam
ABCAAB18575-50EA 0 EUR
ABCAAB18575-50
Anti-FSBA Mouse Monoclonal Antibody [clone: 2F5/5]
Anticuerpos
Type: Primary
Antigen: FSBA
Clonality: monoclonal
Clone: 2F5/5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR