Anti-BCL11B Rat monoclonal antibody unconjugated [clone: 25B6]

Proveedor: Abcam
ABCAAB18465-100EA 865 EUR
ABCAAB18465-100
Anti-BCL11B Rat monoclonal antibody unconjugated [clone: 25B6]
Anticuerpos
Type: Primary
Antigen: BCL11B (B-cell CLL/lymphoma 11B (zinc finger protein))
Clonality: monoclonal
Clone: 25B6
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR