Anti-FASLG Hamster polyclonal antibody

ABCAAB18373-500EA 611 EUR
ABCAAB18373-500
Anti-FASLG Hamster polyclonal antibody
Type: Primary
Antigen: FASLG (Fas ligand (TNF superfamily, member 6))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR