Anti-CD4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-241]

Proveedor: Abcam
ABCAAB18280-50EA 550 EUR
ABCAAB18280-50
Anti-CD4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-241]
Anticuerpos
Type: Primary
Antigen: CD4 (CD4 molecule)
Clonality: monoclonal
Clone: MEM-241
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR