Anti-CD1A Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: HI149]

Proveedor: Abcam
ABCAAB18231-50EA 0 EUR
ABCAAB18231-50
Anti-CD1A Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: HI149]
Anticuerpos
Type: Primary
Antigen: CD1A (CD1a molecule)
Clonality: monoclonal
Clone: HI149
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR