Anti-CD34 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 4H11(APG)]

Proveedor: Abcam
ABCAAB18225-50EA 379 EUR
ABCAAB18225-50
Anti-CD34 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 4H11(APG)]
Anticuerpos
Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: monoclonal
Clone: 4H11(APG)
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR