Anti-CD72 Mouse monoclonal antibody Biotin [clone: 3F3]

ABCAAB18223-100EA 341 EUR
ABCAAB18223-100
Anti-CD72 Mouse monoclonal antibody Biotin [clone: 3F3]
Type: Primary
Antigen: CD72 (CD72 molecule)
Clonality: monoclonal
Clone: 3F3
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR