Anti-B lymphocytes Mouse Monoclonal Antibody [clone: UCH-B1]

Proveedor: Abcam
ABCAAB18201-50EA 0 EUR
ABCAAB18201-50
Anti-B lymphocytes Mouse Monoclonal Antibody [clone: UCH-B1]
Anticuerpos
Type: Primary
Antigen: B lymphocytes
Clonality: monoclonal
Clone: UCH-B1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR