Anti-TFAP2B Mouse Monoclonal Antibody [clone: AP2b 1G6/2]

Proveedor: Abcam
ABCAAB18114-50EA 756 EUR
ABCAAB18114-50
Anti-TFAP2B Mouse Monoclonal Antibody [clone: AP2b 1G6/2]
Anticuerpos
Type: Primary
Antigen: TFAP2B (transcription factor AP-2 beta (activating enhancer binding protein 2 beta))
Clonality: monoclonal
Clone: AP2b 1G6/2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR