Anti-L1CAM Mouse monoclonal antibody [clone: SPM275]

ABCAAB18100EA 619 EUR
ABCAAB18100
Anti-L1CAM Mouse monoclonal antibody [clone: SPM275]
Type: Primary
Antigen: L1CAM (L1 cell adhesion molecule)
Clonality: monoclonal
Clone: SPM275
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR