Anti-ACTN Mouse Monoclonal Antibody [clone: 0.T.02]

Proveedor: Abcam
ABCAAB18061-100EA 495 EUR
ABCAAB18061-100
Anti-ACTN Mouse Monoclonal Antibody [clone: 0.T.02]
Anticuerpos
Type: Primary
Antigen: ACTN (actinin, alpha)
Clonality: monoclonal
Clone: 0.T.02
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR