Anti-INTS8 Rabbit Polyclonal Antibody
Proveedor: Abcam
ABCAAB18050-100EA 510 EUR
ABCAAB18050-100
Anti-INTS8 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: INTS8 (integrator complex subunit 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human