Anti-Synaptobrevin Mouse monoclonal antibody [clone: 4H302]

ABCAAB18015-50EA 577 EUR
ABCAAB18015-50
Anti-Synaptobrevin Mouse monoclonal antibody [clone: 4H302]
Type: Primary
Antigen: Synaptobrevin
Clonality: monoclonal
Clone: 4H302
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR