Anti-CXCL1 Rabbit polyclonal antibody

ABCAAB17882-50EA 619 EUR
ABCAAB17882-50
Anti-CXCL1 Rabbit polyclonal antibody
Type: Primary
Antigen: CXCL1 (chemokine (C-X-C motif) ligand 1 (melanoma growth stimulating activity, alpha))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR