Anti-HSPG2 Rat monoclonal antibody [clone: SPM255]

ABCAAB17848-7EA 634 EUR
ABCAAB17848-7
Anti-HSPG2 Rat monoclonal antibody [clone: SPM255]
Type: Primary
Antigen: HSPG2 (heparan sulfate proteoglycan 2)
Clonality: monoclonal
Clone: SPM255
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR