Anti-Milk Fat Globule 1 Mouse Monoclonal Antibody [clone: SPM291]

Proveedor: Abcam
ABCAAB17787-500EA 815 EUR
ABCAAB17787-500
Anti-Milk Fat Globule 1 Mouse Monoclonal Antibody [clone: SPM291]
Anticuerpos
Type: Primary
Antigen: Milk Fat Globule 1
Clonality: Monoclonal
Clone: SPM291
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR