Anti-Tropomyosin Mouse monoclonal antibody [clone: SPM224]

ABCAAB17784-500EA 619 EUR
ABCAAB17784-500
Anti-Tropomyosin Mouse monoclonal antibody [clone: SPM224]
Type: Primary
Antigen: Tropomyosin
Clonality: monoclonal
Clone: SPM224
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR