Anti-IME1 Chicken polyclonal antibody

ABCAAB17711-50EA 588 EUR
ABCAAB17711-50
Anti-IME1 Chicken polyclonal antibody
Type: Primary
Antigen: IME1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR