Anti-LIPE Chicken polyclonal antibody

ABCAAB17652-250EA 634 EUR
ABCAAB17652-250
Anti-LIPE Chicken polyclonal antibody
Type: Primary
Antigen: LIPE (lipase, hormone-sensitive)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR