Anti-AKT1 Chicken polyclonal antibody

ABCAAB17600-50EA 648 EUR
ABCAAB17600-50
Anti-AKT1 Chicken polyclonal antibody
Type: Primary
Antigen: AKT1 (v-akt murine thymoma viral oncogene homolog 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR