Anti-Phosphoserine/threonine Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB17464-50EA 585 EUR
ABCAAB17464-50
Anti-Phosphoserine/threonine Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Phosphoserine/threonine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR