Anti-APP Mouse Monoclonal Antibody [clone: 4H308]

ABCAAB17420-50EA 611 EUR
ABCAAB17420-50
Anti-APP Mouse Monoclonal Antibody [clone: 4H308]
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: monoclonal
Clone: 4H308
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR