Anti-HIST4H4 Rabbit polyclonal antibody unconjugated

Proveedor: Abcam
ABCAAB17343-100EA 0 EUR
ABCAAB17343-100
Anti-HIST4H4 Rabbit polyclonal antibody unconjugated
Anticuerpos
Type: Primary
Antigen: HIST4H4 (histone cluster 4, H4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR