Anti-Phosphotyrosine Rabbit Polyclonal Antibody
Proveedor: Abcam
ABCAAB17302-50EA 897 EUR
ABCAAB17302-50
Anti-Phosphotyrosine Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: Phosphotyrosine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now