Anti-APP Mouse monoclonal antibody [clone: 19B8]

ABCAAB17250-100EA 577 EUR
ABCAAB17250-100
Anti-APP Mouse monoclonal antibody [clone: 19B8]
Type: Primary
Antigen: APP (amyloid beta (A4) precursor protein)
Clonality: monoclonal
Clone: 19B8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR