Anti-AFP Mouse monoclonal antibody [clone: C3]

ABCAAB17240-500EA 611 EUR
ABCAAB17240-500
Anti-AFP Mouse monoclonal antibody [clone: C3]
Type: Primary
Antigen: AFP (alpha-fetoprotein)
Clonality: monoclonal
Clone: C3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR