Anti-NCAM1 Mouse Monoclonal Antibody [clone: 1213C3.D5]

Proveedor: Abcam
ABCAAB17235-6EA 787 EUR
ABCAAB17235-6
Anti-NCAM1 Mouse Monoclonal Antibody [clone: 1213C3.D5]
Anticuerpos
Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: monoclonal
Clone: 1213C3.D5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR