Anti-SLPI Mouse Monoclonal Antibody (Biotin) [clone: 31]

Proveedor: Abcam
ABCAAB17158-250EA 0 EUR
ABCAAB17158-250
Anti-SLPI Mouse Monoclonal Antibody (Biotin) [clone: 31]
Anticuerpos
Type: Primary
Antigen: SLPI (secretory leukocyte peptidase inhibitor)
Clonality: monoclonal
Clone: 31
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR