Anti-KRT18 Mouse monoclonal antibody [clone: CY-90]

ABCAAB17151-1EA 611 EUR
ABCAAB17151-1
Anti-KRT18 Mouse monoclonal antibody [clone: CY-90]
Type: Primary
Antigen: KRT18 (keratin 18)
Clonality: monoclonal
Clone: CY-90
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR