Anti-KRT5 Mouse Monoclonal Antibody [clone: XM26]

Proveedor: Abcam
ABCAAB17130-500EA 0 EUR
ABCAAB17130-500
Anti-KRT5 Mouse Monoclonal Antibody [clone: XM26]
Anticuerpos
Type: Primary
Antigen: KRT5 (keratin 5)
Clonality: monoclonal
Clone: XM26
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR