Anti-SAST124 Mouse monoclonal antibody [clone: 21]

ABCAAB17054-50EA 588 EUR
ABCAAB17054-50
Anti-SAST124 Mouse monoclonal antibody [clone: 21]
Type: Primary
Antigen: SAST124
Clonality: monoclonal
Clone: 21
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR