Anti-TNFRSF1B Mouse Monoclonal Antibody (Biotin) [clone: MR2-1]

Proveedor: Abcam
ABCAAB16920-250EA 739 EUR
ABCAAB16920-250
Anti-TNFRSF1B Mouse Monoclonal Antibody (Biotin) [clone: MR2-1]
Anticuerpos
Type: Primary
Antigen: TNFRSF1B (tumor necrosis factor receptor superfamily, member 1B)
Clonality: monoclonal
Clone: MR2-1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR