Anti-BAX Mouse monoclonal antibody [clone: ID3]

ABCAAB16910-50EA 577 EUR
ABCAAB16910-50
Anti-BAX Mouse monoclonal antibody [clone: ID3]
Type: Primary
Antigen: BAX (BCL2-associated X protein)
Clonality: monoclonal
Clone: ID3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR