Anti-c Abl Mouse monoclonal antibody [clone: 24-21]

ABCAAB16905-100EA 588 EUR
ABCAAB16905-100
Anti-c Abl Mouse monoclonal antibody [clone: 24-21]
Type: Primary
Antigen: c Abl
Clonality: monoclonal
Clone: 24-21
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR