Anti-PDGFRB Rabbit Polyclonal Antibody

Proveedor: Abcam
ABCAAB16863-50EA 0 EUR
ABCAAB16863-50
Anti-PDGFRB Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: PDGFRB (platelet-derived growth factor receptor, beta polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR